Learn More
Benzophenone-13C, TRC
Product Code. 30005476
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005476 5 mg 5mg
30012666 50 mg 50mg
30010078 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005476 Supplier Toronto Research Chemicals Supplier No. TRCB4280035MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32488-48-5
Molecular Formula 13C C12 H10 O
Molecular Weight (g/mol) 183.21
Synonym Methanone-13C, diphenyl- (9CI, ACI), Diphenylmethanone-13C (ACI), Benzophenone-carbonyl-13C, Carbonyl-13C-benzophenone, Benzophenone-13C1 (carbonyl-13C), Benzophenone-13C (carbonyl-13C)
IUPAC Name diphenyl(1^{13}C)methanone
SMILES O=[13C](c1ccccc1)c2ccccc2
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i13+1
Formula Weight 183.0765
Chemical Name or Material Benzophenone-13C