Learn More
Benzoic Anhydride (>90%), TRC
Product Code. 30135197
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135197 500 g 500g
30137821 100 g 100g
30005774 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135197 Supplier Toronto Research Chemicals Supplier No. TRCB203920500G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93-97-0
Formula Weight 226.063
InChI Formula InChI=1S/C14H10O3/c15-13(11-7-3-1-4-8-11)17-14(16)12-9-5-2-6-10-12/h1-10H
IUPAC Name benzoyl benzoate
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
SMILES O=C(OC(=O)c1ccccc1)c2ccccc2
Synonym Benzoic Acid 1,1'-Anhydride, Benzoic Acid Anhydride, Benzoyl Anhydride, Benzoyl Benzoate, Bis(phenylcarbonyl)ether, NSC 37116;
Chemical Name or Material Benzoic Anhydride
Specifications
Quantity 500 g