missing translation for 'onlineSavingsMsg'
Learn More

Benzetimide hydrochloride, MedChemExpress

Product Code. p-200099340
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488708 10 mg 10mg
16404617 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488708 Supplier Medchem Express Supplier No. HYB1547A
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Benzetimide hydrochloride is a muscarinic acetylcholine receptor antagonist.

Benzetimide hydrochloride is a muscarinic acetylcholine receptor antagonist.

Specifications

Chemical Name or Material Benzetimide hydrochloride
CAS 5633-14-7
Color White
Purity Grade Notes Research
Molecular Formula C23H27ClN2O2
Quantity 10 mg
Synonym R4929
Solubility Information DMSO : ≥ 32 mg/mL (80.21 mM) ∣H2O : 16.67 mg/mL (41.79 mM; Need ultrasonic)
SMILES O=C(C(C1CCN(CC2=CC=CC=C2)CC1)(C3=CC=CC=C3)CC4)NC4=O.[H]Cl
Molecular Weight (g/mol) 398.93
Formula Weight 398.93
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.