missing translation for 'onlineSavingsMsg'
Learn More

Bentamapimod, MedChemExpress

Product Code. 16418757
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
2g
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16418757

Brand: Medchem Express HY14761

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bentamapimod (AS 602801) is an ATP-competitive JNK inhibitor with IC50 of 80 nM, 90 nM, and 230 nM for JNK1, JNK2, and JNK3, respectively.

Bentamapimod (AS 602801) is an ATP-competitive JNK inhibitor with IC50 of 80 nM, 90 nM, and 230 nM for JNK1, JNK2, and JNK3, respectively.

Specifications

Chemical Name or Material Bentamapimod
Physical Form Solid
CAS 848344-36-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H23N5O2S
For Use With (Application) Cancer-Kinase/protease
Synonym AS 602801
Solubility Information DMSO : 14.29 mg/mL (31.23 mM; Need ultrasonic)
SMILES N#CC(C1=NC(OCC2=CC=C(C=C2)CN3CCOCC3)=NC=C1)C4=NC5=CC=CC=C5S4
Molecular Weight (g/mol) 457.55
Formula Weight 457.55
Percent Purity 97.8%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.