Learn More
Bentamapimod, MedChemExpress
Product Code. 16491746
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
2g
500mg
50mg
5mg
Product Code. 16491746
Brand: Medchem Express HY14761
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Bentamapimod (AS 602801) is an ATP-competitive JNK inhibitor with IC50 of 80 nM, 90 nM, and 230 nM for JNK1, JNK2, and JNK3, respectively.
Bentamapimod (AS 602801) is an ATP-competitive JNK inhibitor with IC50 of 80 nM, 90 nM, and 230 nM for JNK1, JNK2, and JNK3, respectively.
Specifications
Chemical Name or Material Bentamapimod
Physical Form Solid
CAS 848344-36-5
Color Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C25H23N5O2S
For Use With (Application) Cancer-Kinase/protease
Synonym AS 602801
Solubility Information DMSO : 14.29 mg/mL (31.23 mM; Need ultrasonic)
SMILES N#CC(C1=NC(OCC2=CC=C(C=C2)CN3CCOCC3)=NC=C1)C4=NC5=CC=CC=C5S4
Molecular Weight (g/mol) 457.55
Formula Weight 457.55
Percent Purity 97.8%
Grade Research