Learn More
Lenz Laborglasintrumente Bends 90°
Click to view available options
Standard Taper Joint No.:
NS 19/26
Unit Size:
Each
Description
- With two NS cones
- Bent (angle: 90°)
- Made of DURAN™ tubing
Specifications
Specifications
| Includes | Two NS cones |
| Material | Duran Glass |
| Standard Taper Joint No. | NS 19/26 |