Learn More
Bemcentinib, MedChemExpress
Product Code. 16438917
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16438917
Brand: Medchem Express HY15150
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Bemcentinib (R428) is a potent and selective inhibitor of Axl with an IC50 of 14 nM.
Bemcentinib (R428) is a potent and selective inhibitor of Axl with an IC50 of 14 nM.
Specifications
Chemical Name or Material Bemcentinib
Physical Form Solid
CAS 1037624-75-1
Color Pink
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C30H34N8
For Use With (Application) Cancer-Kinase/protease
Synonym R428 BGB324
Solubility Information DMSO : 25 mg/mL (49.34 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1=NC(NC2=CC(CC[C@@H](N3CCCC3)CC4)=C4C=C2)=NN1C(N=N5)=CC6=C5C7=CC=CC=C7CCC6
Molecular Weight (g/mol) 506.64
Formula Weight 506.64
Percent Purity 95.02%
Grade Research