Learn More
Bel Art™ Switch-Grid™ Test Tube Rack Grids
Product Code. 13412469 Shop All SP Scienceware Products
Click to view available options
Holds:
Tubes (10 to 13 mm)
Tubes (13 to 16 mm)
Tubes (16 to 20 mm)
Tubes (20 to 25 mm)
Tubes (25 to 30 mm)
Array:
3 x 2
4 x 3
5 x 4
6 x 5
7 x 6
Positions:
12
24
40
60
84
Color:
Blue
Fuchsia
Green
Orange
Yellow
Unit Size:
Pack of 4
Product Code. 13412469
Brand: Bel Art™ F187451300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replacement grids
Replacement grids
  • Chemical resistant polyoxymethylene plastic
  • Can be cleaned with mild detergent, ethanol or isopropyl alcohol
  • Autoclavable at121°C (250°F)
Specifications
Product Type Grid Set
Array 7 x 6
Autoclavable Autoclavable
Color Orange
Holds Tubes (10 to 13 mm)
Material Plastic
Positions 84
Shape Rectangular, Rounded Corners
Shelves None
Stackable Yes
Sterility Non-sterile
Length (Metric) 133 mm
Width (Metric) 102 mm
Height (Metric) 5.6 mm