Learn More
Bel-Art™ Varipet™ Pipettor
Product Code. 10374415 Shop All SP Scienceware Products
Click to view available options
Compatible Tips:
Luer Lock Tip
Unit Size:
Each
Product Code. 10374415
Brand: Bel-Art™ F378951100
Only null left
Add to basket
Add to basket
Self-filling pipetting aid designed for speed, ease, and convenience in a dependable, positive displacement dispenser
  • Polypropylene handle to permit extended use without fatigue.
  • PTFE plunger assures smooth, consistent operation in repetitive tasks.
  • Dispensing volume can be adjusted by turning the knob at the base of the unit.
  • Accuracy is ±0.5%.
  • Teflon™ Tip.
  • With chemical resistance because the fluids only contact the borosilicate glass and Teflon.
  • Supplied with chrome-plated steel Luer lock fitting, complete with removable cannula for general use.
Specifications
Accuracy ±0.5%
Compatible Tips Luer Lock Tip
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color White
Disposable Reusable
For Use With Microplate
Grip Type 2-Ring Handle
Material Polypropylene
Model Vari-pet
Number of Channels 1
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 10 mL
Includes Polypropylene handle, PTFE plunger, Borosilicate Glass syringe, Chrome plated Steel luer lock fitting
Increments 0.2 mL
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato