Learn More
Bel Art™ Swing Jaw™ Tubing Clamps
Product Code. p-30731 Shop All SP Scienceware Products
Click to view available options
Diameter (Metric) Outer Tubing:
10 mm
15 mm
20 mm
Unit Size:
Pack of 3
Product Code. 10478412
Brand: Bel Art™ H182200000
Only null left
Add to basket
Add to basket
Provide nonslip closures for flexible tubing
  • Made of zinc-plated steel
  • Knurled adjusting nut bears on a swinging jaw which permits full opening without removal
  • Removable from tubing without disconnecting apparatus
  • Smooth bearing surfaces will not damage tubing; tubing cannot slip out of clamp
  • Strong enough to use with heavy wall vacuum lines
Specifications
Type Tubing Clamp
Material Zinc-plated Steel
For Use With (Equipment) 10 mm (3/8 in.) O.D. flexible tubing
Diameter (English) Outer Tubing 3/8 in.
Diameter (Metric) Outer Tubing 10 mm
Color Silver
Jaw Type Swing