missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Made of zinc-plated steel
- Knurled adjusting nut bears on a swinging jaw which permits full opening without removal
- Removable from tubing without disconnecting apparatus
- Smooth bearing surfaces will not damage tubing; tubing cannot slip out of clamp
- Strong enough to use with heavy wall vacuum lines
Specifications
Specifications
| Type | Tubing Clamp |
| Material | Zinc-plated Steel |
| For Use With (Equipment) | 20 mm (3/4 in.) O.D. flexible tubing |
| Diameter (English) Outer Tubing | 3/4 in. |
| Diameter (Metric) Outer Tubing | 20 mm |
| Color | Silver |
| Jaw Type | Swing |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?