Learn More
Bel-Art™ Reducing Quick-Disconnects
Product Code. p-200738 Shop All SP Scienceware Products
Click to view available options
Diameter (English) Inner:
0.25 to 0.375 in.
For Use With:
6 to 8mm
Unit Size:
Pack of 12
Product Code. 15595604
Brand: Bel-Art™ H197280000
Only null left
Add to basket
Add to basket
Each consists of two barbed polyethylene connectors which assemble tightly together with a male-female center taper
  • Interchangeable male or female polyethylene connections.
  • All sizes are interchangeable with others to make reducing combinations.
  • For use in connecting and disconnecting vacuum lines and other tubing assemblies.
  • Operate at temperatures up to 80°C (176°F).
Specifications
Autoclavable Not Autoclavable
Diameter (English) Inner 0.25 to 0.375 in.
Material Polyethylene
For Use With 6 to 8mm
Includes Two barbed connectors, one male and one female
Color White
Product Type Quick DisConnect