Learn More
Bel-Art™ Labels for Protective Labeling Tape System
Shop All SP Scienceware Products
Click to view available options
Width (English):
1 in.
1.5 in.
Length (English):
1.5 in.
3 in.
Unit Size:
Pack of 1000
Description
- White paper labels accept any kind of marker; perfect for use under the clear tape to label labware, bottles or samples
Specifications
Specifications
| Adhesive Type | Non-adhesive |
| Color | White |
| Display | Consignment |
| Finish | Matte |
| Format | Label |
| Material | Paper |
| Printable Sides | Double-side Printable |
| Print Technology | Ink Jet |
| Product Type | Label |
| Sterility | Non-sterile |
| Show More |