Description
- Positive-actionclamp opens by squeezing handles together
- Naturally clamps shut to stop flow
- Made from chemical- and stain-resistant Delrin™ plastic and stainless steel
Specifications
Specifications
| Quantity | 3 |
| Type | Spring Style Tubing Clamp |
| Material | Acetal Plastic |
| For Use With (Equipment) | All flexible 0.31 in. (0.78 cm) O.D. tubing |
| Color | White |
| Jaw Type | Angled |
| Diameter (Metric) Inside | 8 mm |