Learn More
Bel-Art™ SP Scienceware™ 12-Channel Mini-Tube Strips
Product Code. 10516952 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
1 mL
Unit Size:
Pack of 4800
Product Code. 10516952
Brand: Bel-Art™ F378570000
Please to purchase this item. Need a web account? Register with us today!
Provide an efficient and inexpensive way to perform pipetting operations with multi-channel pipettors of 4, 8, or 12 channels
  • Medical grade polypropylene resin construction
  • Tolerates temperatures down to -40°C
  • Autoclavable at 121°C (250°F) or frozen to -20°C
  • 1mL capacity
  • 1.53 in. (3.9cm) length
  • 0.25 in. (6.2mm) O.D.
  • Features an attachment bridge between tubes which can be easily separated into single tubes by cutting
  • Bulk 400, 12-tube strips
Specifications
Autoclavable Autoclavable
Capacity (Metric) 1 mL
Material Polypropylene
For Use With (Equipment) Multi-channel pipettors of 4, 8, or 12 channels
Packaging Bulk
Diameter (Metric) 6.2 mm
Length (Metric) 39 mm