missing translation for 'onlineSavingsMsg'
Learn More

behr Labor-Technik™ behrotest™ Workstation Equipment

Product Code. 10646331
Click to view available options
Sufficient For:
12 Samples
6 Samples
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10646331

Brand: behr Labor-Technik™ B00218413

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

behr Labor-Technik™ behrotest™ Workstation Equipment is ideal for use with samples of heavy metal digestion with aqua regia.

  • For use with samples of heavy metal digestion with aqua regia

Specifications

For Use With (Application) 6 samples of heavy metal digestion with aqua regia
Sufficient For 6 Samples
Type Behrotest workstation equipment
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.