missing translation for 'onlineSavingsMsg'
Learn More

behr Labor-Technik™ behrotest™ Karlsruher Flask

Product Code. 10157380
Change view
Click to view available options
:
Capacity, Metric:250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10157380 Capacity, Metric:250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10157380 Supplier behr Labor-Technik™ Supplier No. B00217780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Attain precise volumetric measurement. behr Labor-Technik™ behrotest™ Karlsruher Flask is used in a laboratory to hold reagents, make solutions, collect samples and measure chemical solutions.

  • 250 mL with 60mm stopper

Specifications

Capacity (Metric) 250 mL
Height (Metric) 0.17 m
Width (Metric) 0.12 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.