Learn More
Beclomethasone-d5, TRC
Product Code. 30339295
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339295 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30339295 Supplier Toronto Research Chemicals Supplier No. TRCB13100210MG
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Beclomethasone-d5
CAS 1263143-48-1
Purity Grade Notes HPLC
Molecular Formula C22H24D5ClO5
Quantity 10 mg
Synonym (11β,16β)-9-Chloro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione-d5, 9α-Chloro-16β-methylprednisolone-d5, Beclometasone-d5, Becolvent-d5
InChI Formula InChI=1S/C22H29ClO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1/i4D2,9D,11D2
SMILES O[C@H]1C[C@@]2(C)[C@](C[C@H](C)[C@]2(O)C(C([2H])([2H])O)=O)([H])[C@]3([H])CC([2H])([2H])C4=C([2H])C(C=C[C@]4(C)[C@]31Cl)=O
IUPAC Name (8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-4,6,6-trideuterio-17-(2,2-dideuterio-2-hydroxyacetyl)-11,17-dihydroxy-10,13,16-trimethyl-8,11,12,14,15,16-hexahydro-7H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 413.95
Formula Weight 413.2017
Percent Purity >95