Learn More
Tocris Bioscience™ BDY FL Thalidomide
Description
BDY FL Thalidomide is a high-affinity fluorescent probe (Kd = 3.6 nM). It can be used in time-resolved fluorescence resonance energy-transfer (TR-FRET) cereblon binding assays for PROTAC™ Degrader development. This assay is highly sensitive, selective, and stable. Excitation and emission maxima (λ) are 502 and 510 nm, respectively.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 3-(5,5-Difluoro-7,9-dimethyl-5 H-5λ4,6λ4-dipyrrolo[1,2-c:2 ',1 '-f][1,3,2 ]diazaborinin-3-yl)-N-(8-(2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamido)octyl)propenamide |
| CAS | 2740620-18-0 |
| Quantity | 2 mg |
| Target | Fluorescent Probes |
| Molecular Formula | C37H43BF2N6O7 |
| Purity | 0.95 |