missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Megro™ BD Vacutainer™ Plus 6mL Serum Tubes
Click to view available options
Capacity (Metric):
6 mL
Closure Color:
Red
Unit Size:
Pack of 100
Description
- Increased silica act
- Manufactured from PET
- 100 x 13mm
- Red plug
- With transparent label
Specifications
Specifications
| Product Type | Serum Tube |
| Material | PET |
| Closure Type | Plug |
| Capacity (Metric) | 6 mL |
| Closure Color | Red |
| Includes | Transparent label |
| Length (Metric) | 13 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction