Learn More
BD OptEIA™ Assay Diluent
Product Code. 10617325
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Product Code. 10617325
Brand: BD 555213
Please to purchase this item. Need a web account? Register with us today!
A buffered protein solution containing fetal bovine serum that is intended for use in BD OptEIA ELISA Sets
  • Serves as a blocking agent for ELISA plates
  • Serves as a diluent for ELISA standards/samples
  • Used during the preparation of the Working Detector described in specific BD OptEIA ELISA Set protocols
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Immunoreagent Assay Diluent
For Use With (Application) ELISA
Quantity 500 mL
compliance-icons
Product Identifier
  • assay dilutent
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one and 2-methyl-4-isothiazolin-3-one