missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Mycoflask™, Pour Tubes and Ampuled Culture Media: Orange Serum Broth Concentrate
Specifications
Specifications
| Certifications/Compliance | CE, COMPF |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Orange Serum Broth |
| Product Type | Microbiology Media |
| Formulation | Orange Serum 100mL/L, Yeast extract 30g/L, Pancreatic digest of Casein 100g/L, Dextrose 40g/L, Dipotassium Phosphate 25g/L |
| For Use With (Application) | Cultivation of and enumerating microorganisms associated with spoilage of citrus product |
| Packaging Type | Tube |
| Quantity | 10 x 100 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?