Learn More
BD Human IL-8 ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Description
Includes
Capture antibody, detection antibody, enzyme reagent, standards, instruction/ analysis certificate
To develop enzymelinked immunosorbent assays (ELISA) for natural or recombinant human IL-8 in serum, plasma, and cell culture
supernatants
Enzyme-linked immunosorbent assay
Specifications
Specifications
| Concentration | 3.1 to 200pg/mL |
| Product Type | Human IL-8 ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Human IL-8 in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Human IL-8 monoclonal (capture) Antibody, Biotinylated anti-Human IL-8 monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-8 (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |