Learn More
BD Human GM-CSF ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Description
The OptEIA™ set contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human GM-CSF in serum, plasma, and cell culture supernatants.
- Capture antibody: anti-human GM-CSF monoclonal antibody
- Detection antibody: biotinylated anti-human GM-CSF monoclonal antibody
- Enzyme reagent: Streptavidin-horseradish peroxidase conjugate
- Standards: lyophilized recombinant human GM-CSF
ELISA
Specifications
Specifications
| Product Type | Human GM-CSF ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Human GM-CSF in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Human GM-CSF (capture) Antibody, Biotinylated anti-Human GM-CSF (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human GM-CSF (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | -80°C |