Learn More
BD CFSE
Click to view available options
Quantity:
1 mg
Unit Size:
Each
Description
- Blue laser excitable dye
- Used for flow cytometric monitoring of cell divisions
- The non-fluorescent dye passively diffuses across cell membranes and is cleaved by intracellular esterases within viable cells
Specifications
Specifications
| Quantity | 1 mg |
| For Use With (Application) | Intracellular staining (flow cytometry) |
| Color | Blue |
| Storage Requirements | ≤ -20°C |
| Stain Type | Laser excitable dye |