Learn More
BD Difco™ Fluorescent Antibody Reagents
Click to view available options
Product Type:
FA Bordetella Parapertussis
FA Bordetella Pertussis
Unit Size:
5mL
Description
- Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light
- For use in direct fluorescent identification
Order Info
In 5mL vials.
Specifications
Specifications
| Product Type | FA Bordetella Pertussis |
| Quantity | 5 mL |
| For Use With (Application) | Direct fluorescent antibody technique for the identification of Bordetella pertussis and Bordetella parapertussis |