missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: SFP Agar Base
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, COMPF, ISO |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Shahidi Ferguson Perfringens Agar Base |
| Product Type | Agar |
| Formulation | Yeast extract 5g/L, Proteose Peptone No. 3 7.5g/L, Pancreatic digest of Casein 7.5g/L, Soytone 5g/L, Ferric Ammonium Citrate 1g/L, Sodium Bisulfite 1g/L, Agar 20g/L |
| For Use With (Application) | Detection and enumerating Clostridium perfringens in foods |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?