missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Riboflavin Assay Medium
Description
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
AOAC
Specifications
Specifications
| Certifications/Compliance | AOAC |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Riboflavin Assay Medium |
| Product Type | Riboflavin Assay Medium |
| Formulation | Dextrose 20g/L, Sodium Acetate 15g/L, Vitamin assay Casamino Acids 10g/L, Dipotassium Phosphate 1g/L, Monopotassium Phosphate 1g/L, L-Asparagine 0.6g/L, DL-Tryptophan 0.2g/L, L-Cystine 0.2g/L, Magnesium Sulfate USP 0.4g/L, Adenine Sulfate 20mg/L, Guanine |
| For Use With (Application) | Microbiological assay of Riboflavin |
| Packaging Type | Bottle |
| Quantity | 100 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?