missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Purple Agar Base
Click to view available options
:
Purple Agar Base; 500g
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Purple Agar Base |
| Product Type | Agar |
| Formulation | Proteose Peptone No. 3 1g/L, Beef extract 1g/L, Sodium Chloride 5g/L, Agar 15g/L, Bromcresol Purple 0.02g/L |
| For Use With (Application) | Basal medium to which carbohydrates are added to determine fermentation reactions |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction