missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: MacConkey Sorbitol Agar
Click to view available options
:
MacConkey Sorbitol Agar; 500g
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Certifications
CE, BAM, BS12, CCAM, CMPH2, COMPF, ISO, MCM9, SMWW
Specifications
Specifications
| Certifications/Compliance | CE, BAM, BS12, CCAM, CMPH2, COMPF, ISO, MCM9, SMWW |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | MacConkey Sorbitol Agar |
| Product Type | Agar |
| Formulation | Peptone 15.5g/L, Proteose Peptone 3g/L, D-Sorbitol 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 15g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L |
| For Use With (Application) | Isolation and differentiation of enteric pathogenic escherichia coli Serotype |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction