missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Dubos Broth Base
Click to view available options
:
Dubos Broth Base; 500g
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Dubos Broth Base |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 0.5g/L, Asparagine 2g/L, Polysorbate 80 0.2g/L, Monopotassium Phosphate 1g/L, Disodium Phosphate (anhydrous) 2.5g/L, Ferric Ammonium Citrate 50mg/L, Magnesium Sulfate 10mg/L, Calcium Chloride 0.5mg/L, Zinc Sulfate 0.1mg/L, Copper Sulfate 0.1mg/L |
| For Use With (Application) | Used with Dubos medium Albumin Cultivation of pure cultures of M. tuberculosis |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction