missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Acetate Differential Agar
Click to view available options
:
Acetate Differential Agar; 500g
Specifications
Specifications
| Certifications/Compliance | BAM, COMPF, SMD |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Acetate Differential Agar |
| Product Type | Agar |
| Formulation | Sodium Acetate 2g/L, Magnesium Sulfate 0.1g/L, Sodium Chloride 5g/L, Monoammonium Phosphate 1g/L, Dipotassium Phosphate 1g/L, Dipotassium Phosphate 0.08g/L, Agar 20g/L |
| For Use With (Application) | For differentiation of Shigella species from Escherichia coli |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction