Learn More
BD Cytometric Bead Array (CBA) Mouse CD62L (L-Selectin); Flex Set, Bead E7
Descrizione
Includes
- Mouse soluble CD62L (L-Selectin) Standard
- Mouse soluble CD62L (L-Selectin) PE Detection Reagent
- Mouse soluble CD62L (L-Selectin) Capture Bead E7
Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. Standard component is lyophilized and should be transferred to a 15mL polypropylene tube for reconstitution.
- Mouse reactivity was determined by testing samples with this Felx set
- Must be used in conjunction with CBA Mouse/Rat Soluble Protein Master Buffer Kit, flow cytometer, and FCAP Array Software
Specifica
Specifica
| Antigen | CD62L |
| Cross Reactivity | Mouse |
| Includes | Mouse soluble CD62L (L-Selectin) standard, Mouse soluble CD62L (L-Selectin) PE detection reagent, Mouse soluble CD62L (L-Selectin) capture bead E7 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse CD62L (L-Selectin) flex set (bead E7) |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.