Learn More
BD Cytometric Bead Array (CBA) Human IL-1β Enhanced Sensitivity Flex Set (Bead B4)
Product Code. 15831089
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Product Code. 15831089
Brand: BD 561509
Please to purchase this item. Need a web account? Register with us today!
Includes
  • Capture bead
  • Detection reagent
  • Standard
Lyophilized recombinant Human IL-1β measures soluble analytes present in very low concentrations
  • Set is a bead-based immunoassay capable of measuring human interleukin-1β (IL-1β) in serum, plasma and cell culture supernatant samples
  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Includes 100 tests
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Antigen IL-1beta
Cross Reactivity Human
Includes 1 Vial capture bead, 1 vial detection reagent and 2 vials standard
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Human interleukin-1beta
Product Type Human IL-1beta enhanced sensitivity flex set (bead B4)
compliance-icons
Product Identifier
  • BD Cytometric Bead Array (CBA) Human IL-1
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT