missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human IFN-γ Flex Set, Bead E7

Product Code. 15869268
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15869268 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869268 Supplier BD Supplier No. 558269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Human IFN-γ Standard; Human IFN-γ PE Detection Reagent; Human IFN-γ Capture Bead E7

Sufficient for 100 tests

Reactivity

  • QC Testing: Human
  • Testing in Development: Nonhuman Primate

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Cross Reactivity Human
Includes Human IFN-γ Standard, Human IFN-γ PE Detection Reagent, Human IFN-γ Capture Bead E7
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human IFN-γ Flex Set (Bead E7)
compliance-icons
Product Identifier
  • BD Cytometric Bead Array (CBA) Human IFN-
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 5-chloro-2-methyl-4-isothiazolin-3-one

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.