Learn More
BD Cytometric Bead Array (CBA) Human Anaphylatoxin Kit
Click to view available options
Quantity:
25 Tests
Unit Size:
Each
Description
- Performance has been optimized for analysis of specific proteins in EDTA plasma and serum samples
- Provides sufficient reagents for the quantitative analysis of 80 samples
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | Anaphylatoxin |
| Cross Reactivity | Human |
| Includes | Reagents for capture and detection of C4a, C3a and C5a, standards, Wash Buffer, assay diluent, cytometer setup reagents |
| Quantity | 25 Tests |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human anaphylatoxin kit |