Learn More
BD BD Biosciences™ Microlance™ Hypodermic Needle 24G, Violet
Product Code. 10295184
Click to view available options
Unit Size:
Pack of 100
Product Code. 10295184
Brand: BD 304100
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Latex free replacement neeedles. BD Biosciences™ Microlance™ Hypodermic Needle 24G, Violet can be connected to a Luer adapter or Luer Lock connector
  • Gauge - 27G
  • Length - 3/4 in. (19mm)
  • External Diameter - 0.4mm
  • Wall - Regular
  • Bevel - Regular
  • Sterile
  • Latex free
  • Colour Code - Violet
  • Quantity - 100
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 0.55 mm