missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ MacConkey II, I Plate™ Prepared Media
Description
Tripticase Soy Agar (TSA II)
- For the visualization of hemolytic reactions produced by many bacterial species
- Selective and differential plating medium for the detection of coliform organisms and enteric pathogens
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Trypticase™ Soy Agar (TSA II), MacConkey II Agar |
| Formulation | Trypticase™ Soy Agar with 5% Sheep Blood (TSA II): Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5% ; MacConkey II Agar: Pancreatic Digest of Gelatin 17.0g/L, Sodium Chloride 5.0g/L, Pancreatic Digest of Casein 1.5g/L, Neutral Red 0.03g/L, Peptic Digest of Animal Tissue 1.5g/L,Crystal Violet 0.001g/L, Lactose 10.0g/L, Agar13.5g/L, Bile Salts 1.5g/L |
| For Use With (Application) | Cultivation of fastidious microorganisms and visualization of hemolytic reactions |
| Packaging Type | Plate |
| Quantity | 20/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?