Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Colistin35
Click to view available options
Quantity:
10 Pack
Unit Size:
Pack of 10
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
| Antimicrobial Agents | Colistin35 |
| Concentration | 10 μg |
| Packaging Type | Disc |
| Quantity | 10 Pack |
Safety and Handling
Product Identifier
- Disc Colistin Cl-10 10 Ea
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : colistin sulphate
For in vitro diagnostic use