Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Colistin35
Product Code. p-4660028
Click to view available options
Quantity:
10 Pack
Unit Size:
Pack of 10
Product Code. 11883881
Brand: BD 231278
Please to purchase this item. Need a web account? Register with us today!
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Colistin35
Concentration 10 μg
Packaging Type Disc
Quantity 10 Pack
compliance-icons
Product Identifier
  • Disc Colistin Cl-10 10 Ea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : colistin sulphate
For in vitro diagnostic use