Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ciprofloxacin31
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
| Antimicrobial Agents | Ciprofloxacin31 |
| Concentration | 5 μg |
| Packaging Type | Disc |
| Quantity | 10 Cartridges (50 Discs/Cartridge) |
For in vitro diagnostic use