Learn More
BD BBL™ Prepared Plated Media: BCYE Agar (for Legionella)
Click to view available options
:
10/Pk.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Black |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | BCYE Agar (for Legionella) |
| Formulation | Yeast extract 10g/L, L-Cysteine HCl 0.4g/L, Ferric Pyrophosphate 0.25g/L, ACES Buffer 10g/L, Charcoal, Activated 2g/L, Alpha-Ketoglutarate 1g/L, Agar 15g/L |
| For Use With (Application) | Primary isolation and cultivation of Legionella pneumophila and other Legionella species from environmental samples and clinical specimens |
| Packaging Type | Plate |
| Quantity | 10/Pk. |