Learn More
BD BBL™ MGIT™ Mycobacteria Growth Indicator Tube: Reagents
Description
- Creates an environment suitable for rapid mycobacterial growth
- Recommended for testing sputum, stool specimens, body fluids and tissue
- The tube uses fluorometric technology to aid in the reading of results
- Under UV light, a positive test emits a vivid orange fluorescent glow at the tube base and at the meniscus; a negative test shows little or no glow
- The Mycobacteria can be removed from the tube for further testing
- To use, supplement tube with MGIT OADC Enrichment and MGIT PANTA™ Antibiotic Mixture, inoculate with the prepared specimen and incubate
- Tubes fit into standard procedures for decontamination and waste disposal
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | 2 to 25°C |
| Media Type | Panta Antibiotic Mixture |
| For Use With (Application) | For the detection and recovery of Mycobacteria, testing Sputum, Stool Specimens, Body Fluids And Tissue |
| Quantity | 6 Vials |