Learn More
Gibco™ Bacto™ Proteose Peptone
Product Code. 16249771
Click to view available options
Quantity:
500 g
10 kg
Unit Size:
10kg
500g
For Research Use Only. All usage must comply with product instructions.
Product Code. 16249771
Brand: Gibco™ 212010
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Gibco™ Bacto™ Proteose Peptone is a highly nutritious, animal-based media used in preparing microbiological culture media and producing bacterial toxins.
  • Bacto Proteose Peptones are enzymatic digests of protein
  • Studies show that no single peptone is the most suitable nitrogen source for every microbiological application
  • These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone.
  • Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins
  • Store at 2° to 25°C in cool, dry conditions
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance LUO, MUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Dehydrated, free-flowing granules
Product Type Bacterial Media
Format Pail
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
pH 6.6–7.6 (1.0% solution at 25°C)
Product Line Bacto
Quantity 10 kg
Source Animal Origin