missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Grade A Agar
Click to view available options
:
Dehydrated Agar; Grade A; 454g
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Concentration | 1.5% |
| Additive | Agar |
| Form | Dehydrated |
| For Use With (Application) | Dehydrated culture media |
| Packaging Type | Bottle |
| Quantity | 454 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction