missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Simmons Citrate Agar

Product Code. 11708143
Change view
Click to view available options
Simmons Citrate Agar; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11708143 Simmons Citrate Agar; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11708143 Supplier BD Supplier No. 211620

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For differentiation of gramnegative bacteria on the basis of citrate utilization

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, CCAM, COMPF, ISO
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Simmons Citrate Agar
Product Type Agar
Formulation Ammonium Dihydrogen Phosphate 1g/L, Dipotassium Phosphate 1g/L, Sodium Chloride 5g/L, Sodium Citrate 2g/L, Magnesium Sulfate 0.2g/L, Agar 15g/L, Bromthymol blue 0.08g/L
For Use With (Application) For differentiation of gram negative bacteria on the basis of citrate utilization
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.