missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: M-PA-C Agar
Specifications
Specifications
| Certifications/Compliance | SMWW |
| Form | Dehydrated |
| Media Type | M-PA-C Agar |
| Product Type | Agar |
| Formulation | Yeast extract 2g/L, L-Lysine HCl 5g/L, Sodium Chloride 5g/L, Xylose 1.25g/L, Sucrose 1.25g/L, Lactose 1.25g/L, Phenol red 0.08g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 5g/L, Magnesium Sulfate 1.5g/L, Kanamycin 8mg/L, Nalidixic Acid 37mg/L |
| For Use With (Application) | Selective recovery of Pseudomonas auruginosa from Water |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?