Learn More
BD BBL™ Dehydrated Culture Media: Lysine Decarboxylase Broth
Click to view available options
:
Lysine Decarboxylase Broth; 500g
Description
Applications:
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
Specifications
| Certifications/Compliance | CE, BAM, CCAM, COMPF, ISO, SMD, SMWW |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Lysine Decarboxylase Broth |
| Product Type | Microbiology Media |
| Formulation | Peptone 5g/L, Yeast extract 3g/L, Dextrose 1g/L, L-Lysine 5g/L, Bromcresol Purple 0.02g/L |
| For Use With (Application) | For differentiating microorganisms based on Lysine Decarboxylation |
| Packaging Type | Bottle |
| Quantity | 500 g |