Learn More
BD-1047 dihydrobromide, MedChemExpress
Product Code. 16487248
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16487248
Brand: Medchem Express HY16996A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress BD-1047 (dihydrobromide) is a selective functional antagonist of sigma-1 receptor, shows antipsychotic activity in animal models predictive of efficacy in schizophrenia.
BD-1047 (dihydrobromide) is a selective functional antagonist of sigma-1 receptor, shows antipsychotic activity in animal models predictive of efficacy in schizophrenia.
Specifications
Chemical Name or Material BD-1047 dihydrobromide
Physical Form Solid
CAS 138356-21-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H22Br2Cl2N2
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : 25 mg/mL (57.20 mM; Need ultrasonic) ∣H2O : 25 mg/mL (57.20 mM; Need ultrasonic)
SMILES CN(C)CCN(CCC1=CC=C(Cl)C(Cl)=C1)C.[H]Br.[H]Br
Molecular Weight (g/mol) 437.04
Formula Weight 437.04
Percent Purity 98.14%
Grade Research