missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BCL2L12 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16607934
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16607934 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16607934 Supplier Invitrogen™ Supplier No. PA5101453

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BCL2L12.

This gene encodes a member of a family of proteins containing a Bcl-2 homology domain 2 (BH2). The encoded protein is an anti-apoptotic factor that acts as an inhibitor of caspases 3 and 7 in the cytoplasm. In the nucleus, it binds to the p53 tumor suppressor protein, preventing its association with target genes. Overexpression of this gene has been detected in a number of different cancers. There is a pseudogene for this gene on chromosome 3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL2L12
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BCL2L12
Gene Accession No. Q9HB09
Gene Alias 2810475P17Rik; 5430429M05Rik; BCL2 like 12; Bcl-2 related proline-rich protein; Bcl2l12; Bcl2-L12; Bcl2-L-12; BCL2-like 12 (proline rich); bcl-2-like protein 12; Bcl-2-related proline-rich protein; Bcl-L12; BPR
Gene Symbols BCL2L12
Host Species Rabbit
Immunogen A synthesized peptide derived from human BCL2L12(Accession Q9HB09), corresponding to amino acid residues T97-A147.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75736, 83596
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.