missing translation for 'onlineSavingsMsg'
Learn More

BCL2L11 Monoclonal Antibody (OTI2H5), TrueMAB™, OriGene

Product Code. 17516884
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17516884 30 μL 30µL
17596874 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17516884 Supplier Origene Technologies Supplier No. TA812441S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCL2L11 Monoclonal antibody specifically detects BCL2L11 in Human samples. It is validated for Western Blot

Specifications

Antigen BCL2L11
Applications Western Blot
Classification Monoclonal
Clone OTI2H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GPC1
Gene Accession No. P35052
Gene Alias 1500006F24Rik; BAM; Bcl2 interacting mediator of cell death; bcl-2 interacting mediator of cell death; bcl-2 interacting protein Bim; BCL2 like 11; Bcl-2 like 11; Bcl-2 related apoptotic gene product BimL; bcl2-interacting mediator of cell death; BCL2L11; Bcl2-L-11; BCL2-like 11 (apoptosis facilitator); bcl-2-like protein 11; bcl-2-like protein 11-like; bcl-2-related ovarian death agonist; bcl-2-related ovarian death protein; BIM; Bim EL; Bim-A; BIM-alpha6; BIM-beta6; BIM-beta7; BIMEL; BimEL protein; BimL; BOD; OTTHUMP00000197013; OTTHUMP00000204247; Unknown (protein for MGC:134444)
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 24-530 of human GPC1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2817
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.